Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Autoclavable PE Biohazard Bags, 4 mil
Supplier: VWR
VWR®
11215-826 11215-823 11215-822 11215-824
11215-822CS 395.88 USD InStock
11215-822 11215-823 11215-824 11215-826
VWR® Autoclavable PE Biohazard Bags, 4 mil
Bags Waste Disposal Bags
These heavy-duty bags with extra puncture resistance and high impact strength are constructed of high-molecular, high-density 4 mm polyethylene plastic.
  • Bright red color for quick identification
  • Clearly labeled with biohazard warnings
Autoclavable up to 121 °C (250 °F). Ideal for sterilizing animal cages. Each package includes 20.3 cm (8") twist ties. Bags meet many state and local regulations.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty