Would you like to log in again?
SP Bel-Art Cleanware™ Autoclavable Waste Bags, Bel-Art Products, a part of SP
Supplier: Bel-Art
SCIENCEWARE®
F13174-1008 F13174-1012
11215-853PK 31.97 USD InStock
11215-853 11215-855
SP Bel-Art Cleanware™ Autoclavable Waste Bags, Bel-Art Products, a part of SP
Bags Waste Disposal Bags
Disposable waste bags free bench or hood space when hung from walls, cabinets, or virtually anywhere.
Simply peel off the backing on the adhesive strip across the top and hang close to your work area. The back panel is longer than the front for easy opening. Made of 1 mil thick, high-molecular weight, high-density (HMHD) polyethylene, which is steam autoclavable at 121 °C (250 °F). Conforms to ASTM 1922 Tear Resistance and ASTM 1709 Dart Impact Tests. Maximum weight of contents should not exceed 1.4 kg (3 lbs.).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty