Would you like to log in again?
SP Bel-Art Biohazard Disposal Bags, Bel-Art Products, a part of SP
Supplier: Bel-Art
SCIENCEWARE®
F13164-1923 F13164-3138 F13164-2535 F13164-3848 F13164-1419
47750-754PK 529.89 USD InStock
47750-754 11215-860 47750-748 11215-862 47750-752
SP Bel-Art Biohazard Disposal Bags, Bel-Art Products, a part of SP
Bags Waste Disposal Bags
Bags feature a pre-printed sterilization indicator patch that darkens when autoclaved.
  • Pre-printed sterilization indicator patch changes color with autoclaving
  • Prominently displayed biohazard warning symbol
High temperature steam autoclavable to 135 °C (275 °F).
Standard bags are made from virgin-grade polypropylene that meets or exceeds the Dart Impact Resistance Test requirement of 165 g per ASTM D1709-98 and the ASTM 1922 Tear Resistance Tests. Super Biohazard Disposal Bags are made from a blended polymer that offers maximum protection against dangerous tears and leaks.
Labeling displays standard biohazard warning symbol and precautionary protocols in English, French, German, and Spanish.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty