Would you like to log in again?
SP Bel-Art Silicone Rubber Loops, Bel-Art Products, a part of SP
Supplier: Bel-Art
F20308-0000
11215-974EA 92.08 USD InStock
11215-974
SP Bel-Art Silicone Rubber Loops, Bel-Art Products, a part of SP
Ties
Use these adjustable “rubber bands” for bundling, binding, color coding, securing columns and burettes, controlling flexible tubing, keeping foil covers on canisters, and securing dialysis or biohazard bags.
  • Adjustable silicone rubber construction
  • Tension-control function
  • Remains flexible in autoclave or freezer
They remain flexible from –46 to 121˚ C (–50 to 250 ˚F) for use in the autoclave or freezer. Small, tension-control band slides for a loop range of 5 mm (0.2") to 100 mm (4") in diameter. Includes 10 of each color, red, blue and yellow.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty