Would you like to log in again?
SP Bel-Art Odo-Clave® Autoclave Deodorant Pads, Bel-Art Products, a part of SP
Supplier: Bel-Art
Odo-Clave®
H13199-0000 H13198-0001 H13198-0002 H13198-0003 H13198-0000
11215-880CS 2280.79 USD InStock
11215-880 39988-000 11215-983 11215-985 11215-987
SP Bel-Art Odo-Clave® Autoclave Deodorant Pads, Bel-Art Products, a part of SP
Deodorants and Air Fresheners
Heat-released deodorant pads neutralize autoclave odors with one of four pleasant scents.
  • Effectively neutralize autoclave odors
  • Choice of pleasant scents
  • Long lifespan, especially if stored properly
  • Easy to apply prior to autoclaving
Simply open foil pouch and place pad in bag prior to autoclaving
Also available in triple strength clove that releases three times the deodorant of the standard pads while costing less than two standard pads.
Pads have a three-year lifespan but will last longer if stored in a sealed container.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty