Would you like to log in again?
VWR® Sample Sak™ Sample Bags
Supplier: VWR
VWR® Sample Sak™
740-101 740-100
11216-002CS 458.68 USD InStock
11216-002 11216-000
VWR® Sample Sak™ Sample Bags
Bags Sample Bags
Ideal collection/transport containers for powder and granular samples including grain, seeds, feeds, coffee, powdered soaps, and even damp soil.
  • For solid and semi-solid samples
  • Imprinted writing area
  • Tin tie tabs
Heat-sealed bags are constructed of 50 lb. machine finished white Kraft paper with tin tie tabs. Lined with 1.25 mm polyethylene. Imprinted on each bag is a personalized area for recording sample data and date.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty