Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Whirl-Pak® Sterilized Write-On Bags, Nasco Sampling

Supplier: Nasco

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Whirl-Pak®
B01062 B01447 B01445 B01490WA B01064 B01065 B01297 B01067WA B01446 B01067WA
89132-028CS 460.61 USD InStock
89132-028 35000-678 11216-776 11216-774 11216-500 CA11012-434 11216-772 CA11216-770 89130-776 11216-770
Whirl-Pak® Sterilized Write-On Bags, Nasco Sampling
Bags Sample Bags
Blended, virgin polyethylene film with Write-On strip provides exceptional clarity, superior strength, and sterilized after manufacturing for results you can trust. HACCP, EPA, USDA, FDA approved for food contact.

  • Plus 1 sterilization, post-manufacturing sterilization: Increased sample integrity, perforated closure to maintain sterility
  • Convenient write-on strip for easy labeling
  • Puncture-proof closure tabs: Inner and outer tapes for longevity and stability, wire for easy opening, pouring, and closing
  • Opening pull tabs: For easy opening, non-slip textured Tyvek
  • Continuous perimeter seal: Maximum protection against leaks
  • Tape extends beyond wire to eliminate sharp ends and provide a sturdy grip, wire tabs hold the top open for easy access to add to or remove sample, worry-free, no-leak closure for sample integrity

Bags should not be used at temperatures above 180 °F (82 °C). Bags can be frozen to any temperature, even liquid nitrogen temperatures down to –346 °F (–210 °C) —careful handling is required after freezing. Capacity is measured when the bag is closed and tab is folded over three times. Volume and dimensions are approximate.

Shelf life from sterilization date found on the sterility documentation: 5 years from sterilized date found on your sterility confirmation.

Certifications: Approved by USDA, FDA, EPA and HACCP. Food safe.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty