Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Whirl-Pak® Sterilized Standard Bags, Nasco Sampling

Supplier: Nasco

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Whirl-Pak®
B00990 B01018WA B01018 B01027 B01063 B00736 B01009 B01323 B00679 B01020 B00994WA B00992 B01255WA
66175-612CS 1480.58 USD InStock
66175-612 11216-090 11216-796 11216-422 11216-420 11216-781 11216-089 11216-056 11216-012 11216-373 11216-409 CA11216-796 11216-424
Whirl-Pak® Sterilized Standard Bags, Nasco Sampling
Bags Sample Bags
Blended, virgin polyethylene film provides exceptional clarity, superior strength, and sterilized after manufacturing for results you can trust. HACCP, EPA, USDA, FDA approved for food contact.

  • Plus 1 sterilization, post-manufacturing sterilization: Increased sample integrity, perforated closure to maintain sterility
  • Puncture-proof closure tabs: Inner and outer tapes for longevity and stability, wire for easy opening, pouring, and closing
  • Opening pull tabs: For easy opening, non-slip textured Tyvek
  • Continuous perimeter seal: Maximum protection against leaks
  • Tape extends beyond wire to eliminate sharp ends and provide a sturdy grip, wire tabs hold the top open for easy access to add to or remove sample, worry-free, no-leak closure for sample integrity

Bags should not be used at temperatures above 180 °F (82 °C). Bags can be frozen to any temperature, even liquid nitrogen temperatures down to –346 °F (–210 °C) — careful handling is required after freezing. Capacity is measured when the bag is closed and tab is folded over three times. Volume and dimensions are approximate.

Shelf life from sterilization date found on the sterility documentation: 5 years from sterilized date found on your sterility confirmation.

Certifications: Approved by USDA, FDA, EPA and HACCP. Food safe.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty