Would you like to log in again?
Infecon® Absorbent Strips, Com-Pac
Supplier: Com-Pac
Infecon®
INF-1004
11217-528PK 46.58 USD InStock
11217-528
Infecon® Absorbent Strips, Com-Pac
Mailers and Shippers
Designed for use with Infecon shippers and diagnostic packaging, these absorbent strips provide fast and reliable protection against spills.
Strips have an absorbent capacity of 250mL.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty