Would you like to log in again?
Bitran® Liquid-Tight Specimen Bags, Com-Pac
Supplier: Com-Pac
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Bitran®
4740-S 4754-PE 4756-PE 4742-S 4744-S 4746-S 4741-S 4743-S 4759-PE 4755-PE 4745-S 4758-PE 4753-PE 4747-S 4752-PE
11217-108CS 1870.31 USD InStock
11217-108 11217-548 11217-104 11217-126 11217-128 11217-106 11217-100 11217-122 11217-552 11217-124 11217-102 11217-554 11217-120 11217-550 11217-110
Bitran® Liquid-Tight Specimen Bags, Com-Pac
Bags Sample Bags
Polyethylene and SARANEX® zipper bags are designed for the handling and storage of specimens.
  • Unique manufacturing process eliminates leaks
  • Double track airtight zipper is easy to close
The SARANEX® (Series S) bags provide an exceptional vapor barrier for optimum odor control. Polyethylene (Series PE) bags remain flexible when frozen during storage.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty