Would you like to log in again?
Infecon® 2000 Infectious Substance/Diagnostic Specimen Shipper, Com-Pac
Supplier: Com-Pac
Infecon®
INF-2000
11217-570CS 150.58 USD InStock
11217-570
Infecon® 2000 Infectious Substance/Diagnostic Specimen Shipper, Com-Pac
Mailers and Shippers
Economical, dual-purpose shipper is ideal for shipping test tubes containing infectious substances or diagnostic specimens.
The durable outer box is clearly marked with UN certification markings.
Shipper complies with UN regulations, ICAO/IATA Packing Instruction 602, and applicable DOT regulations.
Ordering information: Includes a 50mL (1.7oz.) capacity absorbent sheet, a liquid-tight, 500mL (16.9oz.) pressure vessel that is able to withstand up to 95kPa of pressure, and a compartmental bubble pouch to separate test tubes and provide cushioning support. Shipper’s declarations and packing instructions are included. Also comes with an infectious substance label and a diagnostic specimen label. Dimensions measurements include 500mL vessel.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty