Would you like to log in again?
Flow Through Cells, METTLER TOLEDO®
Supplier: Mettler-Toledo
51302257
11238-674EA 315 USD InStock
11238-674
Flow Through Cells, METTLER TOLEDO®
Flow Cells
High-quality flow cell especially designed for the InLab electrodes with 12 mm diameter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty