Would you like to log in again?
Ion Selective Half Cell Electrode, METTLER TOLEDO®
Supplier: Mettler-Toledo
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
51340900 51340800 51340700 51340263 51340600 51340300 51340500 51340700 51340300 51340600 51340400 51340900 51341000 51107674 51340400 51340800 51341000 51340500 51340263
11277-350EA 690 USD InStock
CA10530-920 11277-350 11277-330 CA11277-350 11277-340 CA11277-330 11277-332 11277-342 CA11277-340 CA11277-332 11277-334 CA11277-342 11277-344 CA11277-334 11277-336 CA11277-344 CA11277-336 11277-338 CA11277-338
Ion Selective Half Cell Electrode, METTLER TOLEDO®
Electrodes ISE Electrodes
Ion Selective half-cells electrodes consist of two elements, a universal shaft and an ion-specific membrane module.
  • Electrolyte filled
  • Ready to use
  • Tested for wet chemistry
  • Passes stringent requirements
  • Fast response time and requires no maintenance
This module may be exchanged to measure desired ions.
Each ion-selective electrode (ISE) and each membrane module has been tested for wet-chemistry applications. It has to pass the stringent requirements of several direct measurements as well as a typical titration application.
The sodium electrode has a similar construction to a normal pH electrode but uses membrane glass with a higher selectivity for sodium.
Ordering information: Membrane kits available should be ordered separately.
Delivery information: The membrane kit comes with an identification ring (ID ring) and an adapter sleeve which will fit any METTLER TOLEDO titration stand.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty