Would you like to log in again?
InLab® 413 SG 3-in-1 pH Electrodes, METTLER TOLEDO®
Supplier: Mettler-Toledo
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
InLab®
51340288 51340288
11278-372EA 545 USD InStock
CA11278-372 11278-372
InLab® 413 SG 3-in-1 pH Electrodes, METTLER TOLEDO®
Electrodes pH Electrodes
Designed for use with the SevenGo® portable meters.
  • Integrated temperature sensors
  • Fixed cables
  • BNC/Cinch connectors.
  • PEEK shafts
  • Solid XEROLYT® polymer electrolytes
  • No junction required
  • ARGENTHAL™ reference system
Applications include emulsions, suspensions, effluent water, and solutions containing proteins and sulfides. Electrodes feature integrated temperature sensors, fixed cables, and BNC/Cinch connectors. High grade plastic (PEEK) shafts offer a high level of chemical resistance and the solid XEROLYT® polymer electrolytes require no junction. The ARGENTHAL™ reference system improves service life and measurement accuracy at differing sample temperatures.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty