Would you like to log in again?
DG113-SC Combination pH Electrode, Glass Body, METTLER TOLEDO®
Supplier: Mettler-Toledo
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
89632 89632
11278-434EA 585 USD InStock
11278-434 CA46710-000
DG113-SC Combination pH Electrode, Glass Body, METTLER TOLEDO®
Electrodes pH Electrodes
<bold>This combination glass pH electrode with a PTFE movable sleeve junction is ideal for direct pH measurement and acid/base titrations in non-aqueous medium
  • Combination electrode
  • Moveable PTFE sleeve
  • S7 screw head
Electrode features S7 screw head connector is compatible with various cables, so electrode may be used with a wide range of meters and detachable cable.
</bold>
Ordering information: Electrode includes a quality certificate and a serial number for full traceability.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty