Would you like to log in again?
HelixMark® Silicone Stoppers, Freudenberg Medical
Supplier: Freudenberg Medical
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
HelixMark®
70-416-90 70-416-03 70-416-07 70-416-00 70-416-99 70-416-06 70-416-02 70-416-05 70-416-09 70-416-04 70-416-08 70-416-10 70-416-12 70-416-12 70-416-01
11310-718PK 1063.34 USD InStock
11310-718 11310-728 11310-716 11310-726 CA11310-710 11310-736 11310-714 11310-724 11310-734 11310-712 11310-722 11310-732 11310-710 11310-720 11310-730
HelixMark® Silicone Stoppers, Freudenberg Medical
Stoppers Silicone Stoppers
Pharmaceutical-grade, platinum-cured silicone stoppers provide a tight, reliable seal for glass tubes, vials, and flasks
  • Non-toxic and contains no peroxide catalysts
  • Fully autoclavable and sterilizable
  • Full traceability to raw materials
  • Material certification in every package
  • Availabe in sizes 000 to 13
Made without peroxide catalysts, stoppers are composed of a silicone elastomer that meets USP Class VI biocompatibility requirements. Fully traceable to raw materials. Manufactured following Good Manufacturing Practices (GMP) in an FDA-registered medical device facility. Stopper length: 25 mm (0.98").
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty