Would you like to log in again?
KIMAX® Sample Vials, Borosilicate Glass, with Black Phenolic Screw Cap, Kimble Chase
Supplier: DWK Life Sciences
KIMAX®
60951A-40 60951A-12 60951A-8 60951A-4
11310-782CS 204.89 USD InStock
11310-782 11310-780 11310-778 11310-786
KIMAX® Sample Vials, Borosilicate Glass, with Black Phenolic Screw Cap, Kimble Chase
Vials
Amber, screw thread vials with attached black phenolic caps and cone-shaped polyethylene liners.
Manufactured from 51 expansion borosilicate amber glass conforming to USP Type I requirements.
Packaging: Packaged in corrugated trays with partitions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty