Would you like to log in again?
REFILL 1 11 ROLL
Supplier: MICRO ESSENTIAL LABORATORY I
Hydrion Ph 1-11 Paper Refill^ 5 rolls. Wide range ( ph 1-11) paper refills for single roll dispensers. Five rolls of one type paper to each refill case. Two color charts furnished with each refill. Packed in heat sealed aluminum foil and tightly secured in a plastic case.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
REFILL 1 11 ROLL 11310-793 160-1-11 Case of 50 Retrieving
Pack of 5 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty