Would you like to log in again?
White Metal Cap, Pulp/Aluminum Foil Liner, Qorpak®
Supplier: Qorpak
CAP-00451 CAP-06655 CAP-06648 CAP-00439 CAP-06653 CAP-06651 CAP-06650 CAP-00434 CAP-06646 CAP-08684 CAP-06652 CAP-06647 CAP-00418 CAP-06657
11311-582CS 3278.3 USD InStock
11311-582 11310-818 11310-806 16200-414 11310-816 11310-814 89204-923 11310-802 76535-012 11310-812 11310-822 11310-810 76535-004 11310-820
White Metal Cap, Pulp/Aluminum Foil Liner, Qorpak®
Closures Bottle Closures
Pulp/Aluminum Foil lined caps are typically used for food applications as they will not cause odor or taste problems
They can be used for non-acid, non-alkaline products. Great for organic solvents, chrome cleaners, brake fluids and mineral oils.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty