Would you like to log in again?
White Metal Screw Caps, Pulp/Polyethylene Liner, Qorpak®
Supplier: Qorpak
CAP-00419 CAP-05015 CAP-00444 CAP-06659 CAP-06662 CAP-06664 CAP-00458 CAP-00464 CAP-00440 CAP-05016 CAP-00453 CAP-06660 CAP-00435
16087-200CS 773.15 USD InStock
16087-200 11311-152 11311-154 76535-006 11310-804 11311-150 76535-008 89204-937 16198-788 16218-110 89204-934 89204-941 76535-010
White Metal Screw Caps, Pulp/Polyethylene Liner, Qorpak®
Closures Bottle Closures
Pulp/Polyethylene lined metal caps are a great general-purpose closure.
They are ideal for powders and non-acidic food products and consumer products such as jar candles and honey.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty