Would you like to log in again?
F-Style Jugs, High-Density Polyethylene, Qorpak®
Supplier: Qorpak
PLA-03250 PLC-03461 PLC-09289 PLC-08929
89176-160CS 420.67 USD InStock
89176-160 11311-168 11311-630 14003-550
F-Style Jugs, High-Density Polyethylene, Qorpak®
Bottles Carboys
White HDPE F-Style jugs feature a space saving rectangular shape and are ideal for environmental applications.
  • The molded panel allows for easy label application
These HDPE jugs provide an excellent moisture barrier and are impact resistant.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty