Would you like to log in again?
Packer Bottles, PET, Clear, Wide Mouth, Qorpak
Supplier: Qorpak
PLC-07300 PLC-06762 PLC-06836 274632 PLC-08925 PLC-08822 PLA-06570 PLC-07520 PLC-06838 PLC-09008 PLC-06837 PLC-06877 PLC-11297 PLC-07097 PLC-06996 PLC-06594
75880-446CS 41.83 USD InStock
75880-446 11311-636 11311-640 89234-738 82020-058 76487-252 76487-362 76487-250 11311-639 76487-360 76487-256 76487-366 76487-254 76487-364 14003-560 30617-180
Packer Bottles, PET, Clear, Wide Mouth, Qorpak
Bottles Roller Bottles
PET Packer Bottles feature glass like clarity and are durable and rigid.
  • PET packers are stress and impact resistant
  • PET bottles are ideal for oil analysis
They have exceptional gas and moisture barrier properties and a good alcohol and solvent barrier.
Ordering information: Bottle 30617-180 is Vacuum and Ionized. Bottle 89234-738 is Super Cleaned for Oil Analysis.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty