Would you like to log in again?
Glove Box Holders, TrippNT
Supplier: TrippNT
50026 50098 50023
11376-250EA 134.14 USD InStock
11376-250 11376-252 11376-254
Glove Box Holders, TrippNT
Glove Dispensers and Glove Box Holders
Designed to hold one, two, or three dispenser boxes, for easy access to gloves at all times
Top-loading holders are made of white PVC with clear acrylic front for quick identification of glove type and size. Will not bend or warp. Keyhole openings on back of unit allow for easy installation with screws (not included).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty