Would you like to log in again?
Small pH Meter Supplies Organizer, PVC, TrippNT
Supplier: TrippNT
50006
11376-298EA 425.25 USD InStock
11376-298
Small pH Meter Supplies Organizer, PVC, TrippNT
Furniture Shelves
White PVC small pH meter supplies organizer is a great way to store pH meter supplies.
  • Made of chemically resistant PVC
  • The open design and 6" deep shelves make it easy to get larger items in and out
Keep buffers, probes, sample cups, squeeze bottles, waste beakers and wipers all together in one organizer. This workstation is compact, its two shelf design creates high storage capacity with a small footprint. Built-in wiper dispenser and six hole tool holder. The shelves have a 1" safety lip, keeping supplies securely in their place, and each unit has multiple compartments to store supplies.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty