Would you like to log in again?
Finntip™ Pipette Tips, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Finntip™
94060517 9400620 9402160 9402183 9401253 9402010 9400360 9400300 9401113 9400610 9400250 9402073 9400133 9402070 9402163 9400613 94060510 9400303 9402150 9401110 9400327 9401255 9400373 94060520 9400230 9400370 9401030 9402151 9400267 9401250 9402180 9400100 9400310 94060513 9401420 9400263 9401115 9400130 9402030 9400260 9401260 9401410 9402171
53516-020PK 23.72 USD InStock
53516-020 53515-958 14224-024 53515-956 53515-912 14224-022 53515-858 53515-954 14231-116 53515-876 14232-918 53515-950 46620-280 14231-112 11388-532 53515-980 46620-282 11388-534 53516-012 53516-156 53516-178 53516-010 76670-568 53516-154 76637-688 53515-946 53516-150 53515-888 53515-866 53515-944 14233-394 11388-538 53515-886 53515-222 53515-982 53511-901 14232-916 14232-914 53516-148 46620-278 76637-690 53516-166 11388-540
Finntip™ Pipette Tips, Thermo Scientific
Pipette Tips
These high-quality pipette tips are specifically designed to fit Finnpipette® pipettors, but are also compatible with most pipettor brands.
  • Made of high-quality polypropylene
  • Autoclavable
  • Chemical-resistant
Tips in refill trays can be used to quickly refill existing racks.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty