Would you like to log in again?
Digital 'Palette' Refractometers, ATAGO®
Supplier: Atago
3488-PR100SA 3452-PR-201A 3486-PET-109 3405-PR-32A 3478-PR50HO 3480 3442-PR-101A 3462-PR-301A 3477-PR60PA
12000-670EA 1506.04 USD InStock
12000-670 47752-860 12000-672 12000-674 12000-676 97015-910 47752-858 47752-862 47752-864
Digital 'Palette' Refractometers, ATAGO®
Refractometers Handheld Refractometers
ATAGO offers its “Palette” series digital refractometers for a wide range of applications and industries
The PR-32α, PR-101α, PR-201α, and PR-301α feature ELI (External Light Interference) technology, which provides security when performing measurements outside or near a bright window. They also display measurement temperature and allow the user to create up to three user scales, so that no conversion tables are needed.
Model PR-RI is designed for measuring Refractive Index, the measurement will be displayed continuously like an electric newsboard once the sample has been placed.
Ordering information: The PR-32α, PR-101α, PR-201α, and PR-301α are supplied with a two-year manufacturer’s warranty; all other models are supplied with a one-year manufacturer’s warranty. Each refractometer is supplied with a 006P dry battery (9V). PR-301α includes a liquid LG and round sheet. Calibration certificates are available for all models. Contact your Avantor representative for information.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty