Would you like to log in again?
AccuGENE® Running Buffers
Supplier: Lonza
AccuGENE®
50880 50844 50882 51216 50879 50843 50880 50836 50841 50876 50843 50879 50876 50882 51216 50836 50844 50841
12002-122EA 502 USD InStock
12002-122 CA12002-122 CA12001-508 12001-508 CA12001-116 12001-116 CA12001-124 CA12001-024 12001-124 CA12001-122 CA12001-012 CA12001-022 12001-024 12001-122 12001-012 CA12001-120 12001-022 12001-120
AccuGENE® Running Buffers
Buffers Electrophoresis Buffers Running Buffers
AccuGENE™ Molecular Biology Buffers are ready-to-use solutions ideal for a wide range of molecular biology applications. Each reagent is prepared with 18 megOhm water, filtered using a 0.2 micron filter, and filled into sterile bottles.
  • Reliable - Manufactured according to strict quality control standards to ensure lot-to-lot consistency.
  • High Quality - No detectable DNase, RNase, or protease activities unless otherwise noted.
  • Efficient - Ready-made solutions eliminate experiment preparation time.
  • Flexible - Customized solutions are available to meet individual needs
TAE (Tris-Acetate-EDTA) buffer is made from Tris, EDTA and Acetic acid
TBE (Tris hydroxymethyl aminomethane) is made from Tris, EDTA and Boric acid. The addition of borate makes TBA a better conductive medium than TAE so it less prone to overheating and better for longer runs. Borate does inhibit some enzymatic reactions, so it is not often used if the DNA/RNA will be extracted for downstream applications.
MOPS(3-(N-morpholino)propanesulfonic acid) is a buffer introduced by Good et al. in the 1960s and is frequently used as a buffering agent in biology and biochemistry. It has been tested and recommended for polyacrylamide gel electrophoresis and RNA electrophoresis.
Tris glycine native buffer is a Tris buffer with added glycine. It is commonly used in electrophoresis of proteins in their “Native” , non-denatured state thus preserving the molecules' higher-order structure. When SDS is added, it denatures the proteins allowing them to separate by size.
Ordering information: For additional sizes, pH, or concentrations, contact your Avantor sales representative.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty