Would you like to log in again?
AccuGENE™ Molecular Biology Buffers
Supplier: Lonza
AccuGENE™
51203 51202 51203 51202
12001-386EA 132.47 USD InStock
CA12001-386 12001-386 CA12001-384 12001-384
AccuGENE™ Molecular Biology Buffers
Buffers Cloning Buffers
AccuGENE™ Molecular Biology Buffers are ready-to-use solutions ideal for a wide range of molecular biology applications.
  • Reliable - Manufactured according to strict quality control standards to ensure lot-to-lot consistency
  • High Quality - No detectable DNase, RNase, or protease activities unless otherwise noted
  • Efficient - Ready-made solutions eliminate experiment preparation time
  • Flexible - Customized solutions are available to meet individual needs
Each reagent is prepared with 18 megOhm water, filtered using a 0.2 µm filter, and filled into sterile bottles.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty