Would you like to log in again?
AccuGENE™ Buffers
Supplier: Lonza
AccuGENE™
51205 51205 51214 51214
12001-388EA 69.68 USD InStock
CA12001-388 12001-388 CA12001-394 12001-394
AccuGENE™ Buffers
Buffers Electrophoresis Buffers Hybridization Buffers
AccuGENE® buffers are prepared with high quality reagents and use 18 megohm water
  • Reliable - Manufactured according to strict quality control standards to ensure lot-to-lot consistency
  • High Quality - No detectable DNase, RNase, or protease activities unless otherwise noted
  • Efficient - Ready-made solutions eliminate experiment preparation time
  • Flexible - Customized solutions are available to meet individual needs
  • Formulation - 3.0 M NaCl, 0.3 M sodium citrate, pH 7.0
Store all reagents at 18 to 26 °C (64 to 79 °F).
They are manufactured according to strict quality control standards to ensure lot-to-lot consistency.
Ordering information: For additional sizes, pH, or concentrations, contact your Avantor sales representative.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty