Would you like to log in again?
AccuGENE® 1 M Tris HCL Buffer, Lonza
Supplier: Lonza
AccuGENE®
51237 51238 51236 51236 51237 51238
12001-418EA 105.92 USD InStock
CA12001-416 12001-418 CA12001-414 12001-416 12001-414 CA12001-418
AccuGENE® 1 M Tris HCL Buffer, Lonza
Buffers Biological Buffers
AccuGENE™ Molecular Biology Buffers are ready-to-use solutions ideal for a wide range of molecular biology applications.
  • Reliable - Manufactured according to strict quality control standards to ensure lot-to-lot consistency
  • High Quality - No detectable DNase, RNase, or protease activities unless otherwise noted
  • Efficient - Ready-made solutions eliminate experiment preparation time
  • Flexible - Customized solutions are available to meet individual needs
Tris-HCl is a solution containing tris base mixed with hydrocholoric acid. Tris has a pKa of 8.07 at STP and is used in bufferings solutions to prevent large changes in pH level. A staple of electrophoretic and other biological applications, Tris has a useful buffer range of 7 - 9, which coincides with the physiological pH typical of most living organisms. Tris is the 'T' in TAE and TBE Buffers.
Each reagent is prepared with 18 megOhm water, filtered using a 0.2 μm, and filled into sterile bottles.
Ordering information: For additional sizes, pH, or concentrations, contact your Avantor sales representative.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty