Would you like to log in again?
NuSieve® Agarose, Lonza
Supplier: Lonza
NuSieve®
50091 50081 50080 50080 50090 50094 50084 50094 50084 50081 50091 50090
12001-726EA 886.54 USD InStock
CA12001-726 12001-726 CA12001-728 CA12002-106 12001-724 12001-722 12002-106 12001-720 CA12001-720 CA12001-722 CA12001-724 12001-728
NuSieve® Agarose, Lonza
Electrophoresis Gels Agarose and Acrylamide
NuSieve® is a high strength agarose perfect for analytical electrophoresis of small DNA/RNA fragments and In-gel PCR/Transformation/Ligation.
  • All Lonza agarose is quality tested to certify performance
  • DNAse/RNAse free
  • High gel strength
NuSieve™ 3:1 Agarose is a molecular biology grade, standard melting temperature agarose that yields strong gels for fine resolution of small DNA, RNA, and PCR products ≤1 kb
NuSieve™ GTG™ Agarose is a low melting temperature (65º C) agarose that finely resolves DNA fragments, PCR and RT-PCR products ranging from 10 to 1,000 bp. NuSieve™ GTG™ Agarose forms easy-to-handle gels and provides consistent DNA mobility from lot-to-lot and is tested for the presence of proteases, ligases and nucleases. Cloning procedures can be performed directly in remelted NuSieve™ GTG™ Agarose, eliminating costly and time-consuming DNA extraction steps. This agarose is also compatible with reactions carried out in the presence of the remelted agarose gel. NuSieve™ GTG™ Agarose is tested and certified for reliable ligation and transformation of DNA directly in a remelted agarose gel.
These easy-to-handle gels enhance the speed of gel processing and blotting. Performance testing of NuSieve™ 3:1 Agarose ensures fine resolution of DNA fragments up to 1,000 bp, though this agarose is capable of finely resolving DNA fragments ranging from 10 bp up to 1,500 bp. NuSieve™ 3:1 Agarose is designed for analytical electrophoresis.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty