Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
RNA Marker, Lonza
Supplier: Lonza
50575 50575
12001-748EA 590.85 USD InStock
CA12001-748 12001-748
RNA Marker, Lonza
Electrophoresis Reagents Molecular Weight Markers
RNA standards may be denatured with standard procedures, and can be visualized in Northern blots with labeled lambda sequence
Ethidium bromide staining detects 4µg or 4µL on a standard agarose gel; smaller quantities can be detected with GelStar® nucleic acid gel or SYBR® green II gel stain.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty