Would you like to log in again?
SeaKem® Agaroses for Protein Applications
Supplier: Lonza
SeaKem®
50020 50010 50041 50020 50030 50014 50014 50040 50041 50011 50010 50040 50031 50021 50021 50031 50030 50011
12001-878EA 557.73 USD InStock
CA12001-880 CA12001-882 CA12001-872 CA12001-884 CA12001-874 CA12001-886 CA12001-876 CA12001-888 CA12001-878 12001-878 12001-888 12001-876 12001-886 12001-874 12001-884 12001-872 12001-882 12001-880
SeaKem® Agaroses for Protein Applications
Media for Microbiology Dehydrated Media for Microbiology
  • HE Agarose provides enhanced resolution in immunoelectrophoresis (IEP) and serum protein electrophoresis
  • HEEO Agarose is a very high EEO agarose that is useful in applications requiring significant cathodal migration
  • HGT Agarose is a high-gelling temperature (42° C) agarose that exhibits high gel strength and clarity
  • ME Agarose is a moderate EEO agarose that is often used for serum protein electrophoresis
Very large proteins (subunit sizes >200 kDa) are difficult to electrophoretically separate on polyacrylamide gels
SeaKem® HE Agarose is a high EEO agarose that provides enhanced resolution in immunoelectrophoresis (IEP), crossed-IEP, counterimmunoelectrophoresis (CIEP), and serum protein electrophoresis.
SeaKem® HEEO Agarose is a very high EEO agarose that is useful in applications requiring significant cathodal migration, such as IEP of IgG and IgM. SeaKem® HEEO Agarose can also be used to blend with a lower EEO agarose to achieve a specific EEO value.
SeaKem® HGT Agarose is a high-gelling temperature (42° C) agarose that exhibits high gel strength and clarity. SeaKem® HGT Agarose is primarily used in protein electrophoresis applications such as counterimmunoelectrophoresis and crossed immunoelectrophoresis.
Accordingly, Lonza offers a range of agarose types designed for specific, high performance protein applications. SeaKem® agaroses effectively separates high molecule weight proteins and protein complexes, and provides high recovery yields with simple procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty