Would you like to log in again?
MetaPhor® Agarose, Lonza
Supplier: Lonza
50184 50184 50181 50180 50181 50180
12001-920EA 3092.37 USD InStock
CA12001-916 12001-920 CA12001-918 CA12001-920 12001-918 12001-916
MetaPhor® Agarose, Lonza
Media for Microbiology Dehydrated Media for Microbiology
MetaPhor® is an intermediate melting temperature agarose that lets you resolve DNA fragments differing in size by 2%, in the range of 200 bp to 800 bp.
  • All Lonza agarose is quality tested to certify performance
  • High gel strength
  • Standard melting temperature
MetaPhor® Agarose is a high resolution agarose that challenges polyacrylamide. MetaPhor® Agarose is an intermediate melting temperature (75°C) agarose with twice the resolution capabilities of the finest-sieving agarose products. For example, a 200 bp DNA fragment can be separated from a 204 bp fragment. Using fast running protocols DNA differing in size by 1% can be resolved in as little as 1.5 hours in a 20 cm long horizontal or vertical gel format. MetaPhor agarose gels (2% to 4%) approximate the resolution of polyacrylamide gels (4% to 8%). These agarose gels are ideal for resolving AMPFLPs, STRs, and tri- and tetranucleotide repeats. Ideal for small PCR fragments, MetaPhore® is the choice agarose for resolving hard to see bands.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty