Would you like to log in again?
SeaPrep® Agarose, Lonza
Supplier: Lonza
SeaPrep®
50302 50302
12001-922EA 496.42 USD InStock
12001-922 CA12001-922
SeaPrep® Agarose, Lonza
Electrophoresis Gels Agarose and Acrylamide
SeaPrep® is an ultra low melting temperature, low eeo agarose perfect for hybridoma cloning, embedding cells and creating cDNA libraries.
  • All Lonza agarose is quality tested to certify performance
  • DNAse/RNAse free
  • Ultra Low melting temperature
  • Store agarose at 18 to 26 °C (64 to 79 °F)
SeaPrep® Agarose
SeaPrep® Agarose is a specialty 'soft agarose' product with ultralow gelling and melting temperature properties that make it valuable for many techniques involving biomolecules. SeaPrep® Agarose is an excellent medium for hybridoma cloning and produces soft, clear gels. SeaPrep® Agarose can be used to expand cDNA plasmid libraries in a strictly representative fashion, decreasing the possibility that less abundant clones vanish during amplification due to differential rates of replication.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty