Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
ProSieve™ Protein Markers
Supplier: Lonza
ProSieve®
50550 50552 50550 50552 50552
12002-114EA 92.67 USD InStock
CA12001-806 CA12002-114 12002-114 470177-250 12001-806
ProSieve™ Protein Markers
Electrophoresis Reagents Western Blotting Reagents
During electrophoresis, these markers help monitor the efficiency of protein separation and allow accurate sizing of proteins in a sample. During Western blotting, they help confirm transfer has occurred from the gel to the membrane.
  • Fast - No more mixing standards
  • Easy - Simply unwrap and use
The color markers contain nine proteins labeled with fluorescent dyes and are supplied as lyophilized solids containing the buffer salts and detergent found in the typical Laemmli buffer system. The proteins do not contain oligosaccharides that can cause anomalous migration, heterogeneous bands, and inaccurate size estimation.
The masses of the stained proteins are: 10, 15, 20, 25, 40, 50, 80, 125, 190 kDa. Each lot of the colored markers is calibrated against the unstained markers for accuracy.
The masses of the un-stained proteins are: 5, 10, 15, 25, 35, 50, 75, 100, 150, and 225 kDa. The 50 kDa band is double-intensity for easy identification.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty