Would you like to log in again?
IsoGel® Precast Agarose IEF Plates, Lonza
Supplier: Lonza
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
IsoGel®
56015 56018 56024 56015 56024 56018
12002-192EA 661.15 USD InStock
12002-192 CA12001-452 12001-452 CA12002-194 12002-194 CA12002-192
IsoGel® Precast Agarose IEF Plates, Lonza
Electrophoresis Gels Precast Gels
Use these precast gels to analyze antibodies and proteins up to 2000kDa
The proteins can be quickly transferred from gel to membrane, stained in situ, or detected by antibodies within one hour of loading the gel. Each gel is supported on GelBond® film to provide dimensional stability throughout the IEF processing, and the convenient 125x100mm (1/2x3/8") gel size fits most horizontal IEF chambers. Each lot of plates is functionally tested and a Certificate of Analysis is available upon request. Store plates at 2 to 8°C (36 to 46°F) for 12 months from the date of manufacture. Store accessories at 18 to 26°C (64 to 79°F).
Ordering information: Plates come in packs of six. The plate accessory pack (12001-450) comes with enough masks, 125mm wicks, and blotting paper for six plates. The accessory bulk pack (12002-142) comes with 100 each of 125mm wicks and blotting paper.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty