Would you like to log in again?
Lockout Tags, Brady Worldwide®
Supplier: Brady
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
66067 66054 66057 50258 66054 65505 66064 65534 65521 65522 66062 66060 65506 65524 65525 50198 66056 65521 65524 66055 65527 65523 65522 50250 65526 65527 66063 65504 66064 66067 66062 65506 50198 65671 66060 65525 66065 65523 50265 66066 66052 50256 65535 66053 66053 65670 66065 65526 65535 65504 66056 66061 66061 66052 66063 65503 50250 65515 65670 66066 50253 50199 65505 50261 65534 50251 65515 66055 50266 50199 65503
12222-058PK 97.65 USD InStock
CA12222-022 12222-058 12222-014 CA12222-060 12222-012 CA12222-062 12222-018 CA12222-028 CA12222-106 12222-016 CA12222-108 CA12222-024 CA12222-026 CA12222-148 CA12222-104 89007-154 89007-158 89007-190 89007-152 12222-042 CA12222-012 12222-044 12222-000 CA12222-018 12222-006 CA12222-058 CA12222-014 12222-008 CA12222-016 89007-164 89007-160 CA12222-042 12222-152 CA12222-044 12222-074 12222-150 12222-156 12222-078 12222-110 CA12222-006 12222-116 CA12222-008 12222-118 89007-170 89007-174 CA12222-152 12222-064 CA12222-076 CA12222-110 CA12222-078 12222-062 12222-024 CA12222-150 CA12222-074 12222-022 12222-106 CA12222-116 12222-028 12222-104 CA12222-118 12222-148 12222-026 CA12222-156 12222-108 CA89007-154 89007-188 CA89007-152 CA89007-158 89007-180 12222-060
Lockout Tags, Brady Worldwide®
Security Locks
Constructed of durable polyester, these lockout tags exceed OSHA's 50lb
  • Rigid polyester surface is easy to write on and stays legible longer than cardstock
  • 3/8" diameter brass eyelet accepts most padlock shackles
  • Tag size is 5.75"Hx3"W
  • Standard package includes 25 tags and nylon fasteners
pull requirement for tagout devices.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty