Constructed of durable polyester, these lockout tags exceed OSHA's 50lb
- Rigid polyester surface is easy to write on and stays legible longer than cardstock
- 3/8" diameter brass eyelet accepts most padlock shackles
- Tag size is 5.75"Hx3"W
- Standard package includes 25 tags and nylon fasteners
pull requirement for tagout devices.