Would you like to log in again?
Labeled Lockout Hasps, Brady Worldwide®
Supplier: Brady
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
65972 65974 65975 65964 65971 65970 65975 65961 65967 65964 65970 65973 65960 65967 65974 65972 65961 65960 65973 65971
12222-098PK 192.42 USD InStock
12222-098 CA12222-098 12222-096 12222-140 CA12222-088 CA12222-100 12222-102 CA12222-094 12222-100 CA12222-140 12222-088 CA12222-096 CA12222-102 12222-090 CA12222-090 CA12222-080 CA12222-092 12222-094 12222-092 12222-080
Labeled Lockout Hasps, Brady Worldwide®
Security Locks
Constructed from tough 5052 anodized aluminum alloy with tamperproof locking tabs, hasp is impossible to remove while lock is still in place
Front and back of hasp feature a write-on surface that accepts pencil, pen, or marker. Markings can be easily erased allowing for repeated use. Each hasp has six lock holes, 0.8cm (5/16") in diameter. Front legend reads: “DO NOT OPERATE. This lock/tag may only be removed by:” followed by space for writing name, department, and expected completion. Back legend reads: “DANGER. This energy source has been LOCKED OUT. Only the individual who signed the reverse side may remove this lock/tag," followed by space for remarks.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty