Would you like to log in again?
Red Lockout Hasps, Brady Worldwide®
Supplier: Brady
65376 65375 65376 65375
12222-226PK 293.91 USD InStock
CA12222-226 12222-226 CA12222-228 12222-228
Red Lockout Hasps, Brady Worldwide®
Security Locks
Lockout hasps are the answer to safety concerns during group lockout situations
  • Answer to safety concerns during group lockout situations
  • Vinyl-coated, high-tensile steel; plated to resist rust
  • Lock holes are ⅜" diameter to accommodate most standard lock shackles
Hasps are constructed of vinyl-coated, high tensile steel and are plated to resist rust. Lock holes are 3/8" diameter to accommodate most standard lock shackles.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty