Would you like to log in again?
Circuit Breaker Lockouts, Brady®
Supplier: Brady
65321 65965 65397 65329 65396 65396 65966 65397 65321 65966 65329 65688 65965 65688
12222-832PK 145.01 USD InStock
12222-832 CA12222-832 CA12222-756 12222-830 CA12222-834 12222-840 12222-836 12222-758 12222-834 CA12222-840 CA12222-830 12222-756 CA12222-758 CA12222-836
Circuit Breaker Lockouts, Brady®
Security Locks
Made of rugged polypropylene and glass-filled, impact-modified nylon, these devices are designed for various breakers and are available in three styles: snap-on, clamp-on, and oversized
  • Rugged polypropylene and glass-filled, impact-modified nylon
  • Accepts lock shackles up to 7 mm (9/32") in diameter and are easily installed
  • Snap-On lockout snaps into place
  • Clamp-On and oversized lockouts do not require screwdriver due to the patented thumbscrew design
Lockouts accept lock shackles up to 7 mm (9/32") in diameter and are easily installed. Snap-on lockout simply snaps into place. Clamp-on and oversized lockouts do not require a screwdriver due to the patented thumbscrew design. Simply tighten lockout securely onto switch tongue, pull cover over thumbscrew, and lock cover to prevent clamp from being loosened. Clamp-on and oversized lockouts feature a blade design for a tighter grip with less torque on the thumbscrew.
The snap-on style locks out 120V breakers with holes in the switch tongue. Dual cavities accommodate different size switch tongues. The versatile, clamp-on style locks out 120/277V and 480/600V breakers and works on a wide range of single-pole and internal-trip, multi-pole breakers. The oversized lockout is ideal for 480/600V switches up to 5.7cm (21/4") wide and 2.2 cm (7/8") thick.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty