Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Gate Valve Lockouts, Brady Worldwide®

Supplier: Brady

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

65599 65593 65589 65587 65592 65599 65598 65588 65598 65560 65597 65588 65592 65597 65594 65561 65564 65564 65562 65562 65560 65594 65563 65561 65589 65563 65587 65593
12222-790EA 213.66 USD InStock
12222-790 CA12222-780 CA12222-782 12222-794 12222-792 12222-776 CA12222-788 12222-796 12222-774 CA12222-784 12222-778 CA12222-786 CA89011-760 CA89011-762 12222-780 CA12222-790 CA12222-792 12222-784 12222-782 CA12222-776 12222-788 12222-786 CA12222-778 CA12222-794 CA12222-774 CA12222-796 89011-760 89011-762
Gate Valve Lockouts, Brady Worldwide®
Security Locks
Made of injection-molded polypropylene, the gate valve lockouts are extremely rugged for superior impact and chemical resistance

  • Made of injection-molded polypropylene
  • Extremely rugged for superior impact and chemical resistance
  • Withstand temperatures from –28.9 to 148.9°C (–20 to 300°F)
  • Unique knockout center accommodates OS&Y or "rising stem" valves
  • Lockouts accept locks with shackles up to 9.52mm (⅜") diameter

They can easily withstand temperatures from –28.9 to 148.9°C (–20 to 300°F). Unique knockout center accommodates OS&Y or "rising stem" valves. Lockouts accept locks with shackles up to 9.52mm (⅜") diameter.

Ordering information: Danger labels in English, French, and Spanish are included.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty