Would you like to log in again?
Adjustable Gate Valve Lockout, Brady®
Supplier: Brady
64057
12222-798EA 86.28 USD InStock
12222-798
Adjustable Gate Valve Lockout, Brady®
Security Locks
Versatile, economical lockout adjusts to fit 2.5–16.5cm (1–61/2") diameter valve handles, replacing three conventional gate valve lockouts
Made of durable polypropylene for resistance to extreme temperature changes from –46 to 182°C (–50 to 360°F). Device is compact for easy carrying and storage and features front legend that reads: “DANGER. LOCKED OUT. DO NOT OPERATE.”
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty