Would you like to log in again?
Lockout for Intermediate SMC Models AR3000/NAR3000, Brady Worldwide®
Supplier: Brady
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
64540 64540
12222-828EA 64.39 USD InStock
CA12222-828 12222-828
Lockout for Intermediate SMC Models AR3000/NAR3000, Brady Worldwide®
Security Locks
Lockout enables regulator to be locked in the on, off, or throttled position
Easy installation involves removing snap cap, installing lockout on regulator rod, and snapping replacement cap on regulator. Supplied with lockout, cap, and allen wrench.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty