Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

120 V Snap-On Circuit Breaker Lockout, Brady Worldwide®

Supplier: Brady

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

65387 65387
12222-838EA 13.24 USD InStock
12222-838 CA12222-838
120 V Snap-On Circuit Breaker Lockout, Brady Worldwide®
Security Locks
Designed for use with 120V circuit breakers that have holes in the switch tongue

Dual cavities accommodate different size switch tongues, and the device accepts lock shackles up to 7.1mm (0.281") in diameter. Dimensions are 21Hx8.9Wx55L mm (0.83x0.35x2.15").

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty