Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Polyethylene and Antistatic Polyethylene Bags and Tubing, KNF Clean Room Products

Supplier: KNF
600087-06 600019-28 600094-04 600087-28 600009-72 600009-47 600094-13 600094-08 600087-23 600019-37 600087-25 600009-45 600009-61 600009-88 600087-09 600019-10 600094-17 600019-01 600094-22
12243-062PK 508.98 USD InStock
12243-062 12243-040 12243-116 12243-028 12243-126 12243-104 12243-022 12243-032 12243-120 12243-054 12243-064 12243-086 12243-130 12243-052 12243-074 89089-120 12243-068 12243-134 12243-100
Polyethylene and Antistatic Polyethylene Bags and Tubing, KNF Clean Room Products
Bags Sample Bags
This tough yet flexible film is made of low-density, puncture-resistant polyethylene Ultraclean Film™ for applications requiring Level II (poly) and III (antistatic poly) packaging cleanliness.

  • Thickness: 6 mil
  • Tubing can be easily heat sealed
  • Chemically resistant to most solvents
  • Low water vapor transmission rate

Pink antistatic poly bags and tubing feature good electrical properties.

Surface resitivity: 4.5×1011 Ohm
Dart impact: 350 g

Certifications: FDA approved for use with food.

Caution: Flammable

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty