Would you like to log in again?
Polyethylene and Antistatic Polyethylene Bags and Tubing, KNF Clean Room Products
Supplier: KNF
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
600087-06 600019-28 600094-04 600087-28 600009-72 600009-47 600094-13 600094-08 600087-23 600019-37 600087-25 600009-45 600009-61 600009-88 600087-09 600019-10 600094-17 600019-01 600094-22
12243-062PK 508.98 USD InStock
12243-062 12243-040 12243-116 12243-028 12243-126 12243-104 12243-022 12243-032 12243-120 12243-054 12243-064 12243-086 12243-130 12243-052 12243-074 89089-120 12243-068 12243-134 12243-100
Polyethylene and Antistatic Polyethylene Bags and Tubing, KNF Clean Room Products
Bags Sample Bags
This tough yet flexible film is made of low-density, puncture-resistant polyethylene Ultraclean Film™ for applications requiring Level II (poly) and III (antistatic poly) packaging cleanliness.
  • Thickness: 6 mil
  • Tubing can be easily heat sealed
  • Chemically resistant to most solvents
  • Low water vapor transmission rate
Pink antistatic poly bags and tubing feature good electrical properties.
Certifications: FDA approved for use with food.
Caution: Flammable
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty