Would you like to log in again?
Nylon and Antistatic Nylon Bags and Tubing, KNF Clean Room Products
Supplier: KNF
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
600100-11 600100-06 600027-12 600023-55 600023-62 LB302:1416V 600027-20 600098-22 600098-05 600098-16 600023-30 600098-09 600100-05 600027-30 600027-28 600098-11 600100-09
89004-838CS 823.93 USD InStock
89004-838 12243-182 12243-226 12243-214 12243-208 12243-176 12243-186 12243-174 89089-101 12243-202 12243-224 12243-234 12243-146 12243-212 12243-156 12243-178 12243-232
Nylon and Antistatic Nylon Bags and Tubing, KNF Clean Room Products
Bags Sample Bags
Nylon 6 Ultraclean Film™ material is certified to Level I cleanliness standards and features superior gas barrier properties, and resistance to heat, oil and grease, abrasion, wear, and puncture.
  • Autoclavable
  • Thickness: 2 mil
Film can be used as packaging for electronics, pharmaceuticals, precision parts, and more.
Caution: Flammable.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty