Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Breather Bags, Sheeting and Tubing, KNF

Supplier: KNF

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

600105-07 600076-04 600037-59 600076-09 600076-07 600076-10 600076-08 600037-46 600105-01 600037-14
12243-844CS 1008.57 USD InStock
12243-844 12243-842 12243-836 12243-252 12243-262 12243-838 12243-272 12243-840 12243-246 12243-278
Breather Bags, Sheeting and Tubing, KNF
Bags Sterilization Pouches
Ideal for use in pharmaceutical and medical industries where FDA Class 100 sterilization is required.

Bags are made from two sheets of copolymer polypropylene with a strip of Tyvek® 1059B heat-sealed over the slits on one of the sheets.

When subjected to varying chamber pressures, the breather slits help equalize internal and external pressures, while preventing product from contacting the outer Tyvek® barrier. Breather slits are 10.16 cm (4") wide and located at the center of bag or sheet. When sizes cannot be predetermined, Breather Tubing is also available. Thickness: 3 mil.

Sterilizable and disposable, these products can withstand temperatures up to 128 °C for extended periods and remain exceptionally pliable.

Certifications: Cleanliness level 1.

Packaging: Supplied triple-barrier packaged with nylon inner bag and two polyethylene outer bags.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty